The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109758.1 | NP_001103228.1 | 1 - 2 | 253 | 115 |
NM_007529.2 | NP_031555.2 | 1 - 2 | 253 | 115 | |
GenBank mRNA | CX219984.1 | - | 1 - 2 | 253 | 115 |