Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcan
Entrez Gene ID
12032
Gene Name
brevican
Gene Aliases
Cspg7
Chromosome Location
Chr.3: 87987531 - 88000893 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109758.1 NP_001103228.1 3 - 4 737 98
NM_007529.2 NP_031555.2 3 - 4 737 98
GenBank mRNA AK144405.1 - 3 - 4 537 98
BC052032.1 - 3 - 4 532 98

View Full Product Details