Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcan
Entrez Gene ID
12032
Gene Name
brevican
Gene Aliases
Cspg7
Chromosome Location
Chr.3: 87987531 - 88000893 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007529.2 NP_031555.2 9 - 10 2253 121
GenBank mRNA AK144405.1 - 9 - 10 2054 121
AK163646.1 - 2 - 3 314 121
BC052032.1 - 9 - 10 2048 121
X87096.1 - 9 - 10 2129 121

View Full Product Details