The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176808.5 | NP_789778.1 | 1 - 2 | 113 | 90 |
GenBank mRNA | AK075781.1 | - | 1 - 2 | 91 | |
AK141599.1 | - | 1 - 2 | 71 | 90 |