The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159729.1 | NP_001153201.1 | 2 - 3 | 261 | 67 |
NM_008999.4 | NP_033025.2 | 2 - 3 | 373 | 67 | |
GenBank mRNA | AK016493.1 | - | 2 - 3 | 273 | 67 |
AK045454.1 | - | 2 - 3 | 132 | 67 | |
AK047068.1 | - | 2 - 3 | 374 | 67 | |
AK054079.1 | - | 2 - 3 | 359 | 67 | |
AK082017.1 | - | 2 - 3 | 335 | 67 | |
AK163909.1 | - | 2 - 3 | 382 | 67 | |
AK164057.1 | - | 2 - 3 | 360 | 67 | |
BC025578.1 | - | 2 - 3 | 268 | 67 |