Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slain1
Entrez Gene ID
105439
Gene Name
SLAIN motif family, member 1
Gene Aliases
9630044O09Rik, AA675320, AW742596
Chromosome Location
Chr.14: 103650071 - 103704912 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198014.2 NP_932131.2 4 - 5 1297 63
GenBank mRNA AK036190.1 - 6 - 7 1402 63
BC025223.1 - 2 - 3 451 63
BC079866.1 - 4 - 5 1297 63

View Full Product Details