The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028138.1 | NP_082414.1 | 6 - 7 | 919 | 82 |
GenBank mRNA | AK012806.1 | - | 6 - 7 | 902 | 82 |