Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ky
Entrez Gene ID
16716
Gene Name
kyphoscoliosis peptidase
Gene Aliases
CD1, D9Mgc44e, ky
Chromosome Location
Chr.9: 102505755 - 102546252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024291.3 NP_077253.3 3 - 4 261 121
GenBank mRNA AK047076.1 - 3 - 4 652 121

View Full Product Details