The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026070.3 | NP_080346.1 | 4 - 5 | 469 | 61 |
NM_027487.1 | NP_081763.1 | 4 - 5 | 469 | 61 | |
GenBank mRNA | AK004206.1 | - | 4 - 5 | 384 | 61 |
AK009358.1 | - | 4 - 5 | 365 | 61 | |
AK013835.1 | - | 4 - 5 | 474 | 61 | |
AK017732.1 | - | 4 - 5 | 397 | 61 | |
AK158822.1 | - | 4 - 5 | 470 | 61 | |
BC029256.1 | - | 4 - 5 | 61 |