Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfap2d
Entrez Gene ID
226896
Gene Name
transcription factor AP-2, delta
Gene Aliases
Tcfap2d
Chromosome Location
Chr.1: 19103022 - 19168592 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153154.2 NP_694794.1 7 - 8 1378 67
GenBank mRNA AF421891.1 - 7 - 8 1405 67
AK132112.1 - 7 - 8 1380 67
BC152324.1 - 7 - 8 1327 67
BC152325.1 - 7 - 8 1327 67

View Full Product Details