The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029961.2 | NP_084237.1 | 20 - 21 | 2508 | 91 |
GenBank mRNA | AY766239.1 | - | 20 - 21 | 2472 | 91 |
JQ655148.1 | - | 7 - 8 | 1987 | 91 |