The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198865.1 | NP_942565.1 | 2 - 2 | 2957 | 142 |
GenBank mRNA | AK173062.1 | - | 1 - 1 | 1988 | 142 |