Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif4
Entrez Gene ID
16571
Gene Name
kinesin family member 4
Gene Aliases
AI323435, D330050K22Rik, Kns4
Chromosome Location
Chr.X: 100626065 - 100727271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008446.2 NP_032472.2 17 - 18 2439 144
GenBank mRNA AK135430.1 - 17 - 18 1933 144
AK171938.1 - 17 - 18 2351 144
BC050946.1 - 17 - 18 2377 144
BC054537.1 - 17 - 18 2361 144
D12646.1 - 17 - 18 2766 144

View Full Product Details