Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif4
Entrez Gene ID
16571
Gene Name
kinesin family member 4
Gene Aliases
AI323435, D330050K22Rik, Kns4
Chromosome Location
Chr.X: 100626065 - 100727271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008446.2 NP_032472.2 9 - 10 1544 92
GenBank mRNA AK084837.1 - 9 - 10 1521 92
AK135179.1 - 9 - 10 1492 92
AK135430.1 - 9 - 10 92
AK171938.1 - 9 - 10 1455 92
BC050946.1 - 9 - 10 1482 92
BC054537.1 - 9 - 10 1466 92
D12646.1 - 9 - 10 1871 92

View Full Product Details