Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sycp3
Entrez Gene ID
20962
Gene Name
synaptonemal complex protein 3
Gene Aliases
Cor1, Scp3
Chromosome Location
Chr.10: 88459587 - 88473236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011517.2 NP_035647.2 3 - 4 330 131
GenBank mRNA AK131949.1 - 3 - 4 254 131
BC048483.1 - 3 - 4 253 131
BC089540.1 - 3 - 4 253 131
BC132079.1 - 3 - 4 331 131
BC138509.1 - 3 - 4 331 131
Y08485.1 - 3 - 4 339 131

View Full Product Details