Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plod2
Entrez Gene ID
26432
Gene Name
procollagen lysine, 2-oxoglutarate 5-dioxygenase 2
Gene Aliases
D530025C14Rik, LH2, Plod-2
Chromosome Location
Chr.9: 92542223 - 92608428 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142916.1 NP_001136388.1 18 - 19 2237 80
NM_011961.3 NP_036091.2 17 - 18 2174 80
GenBank mRNA AF080572.1 - 17 - 18 1945 80
AK033363.1 - 18 - 19 2196 80
AK046364.1 - 17 - 18 2178 80
AK157207.1 - 18 - 19 2197 80
BC021352.1 - 17 - 18 2006 80

View Full Product Details