Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plod2
Entrez Gene ID
26432
Gene Name
procollagen lysine, 2-oxoglutarate 5-dioxygenase 2
Gene Aliases
D530025C14Rik, LH2, Plod-2
Chromosome Location
Chr.9: 92542223 - 92608428 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142916.1 NP_001136388.1 7 - 8 1014 91
NM_011961.3 NP_036091.2 7 - 8 1014 91
GenBank mRNA AF080572.1 - 7 - 8 785 91
AK033363.1 - 7 - 8 973 91
AK046364.1 - 7 - 8 1018 91
AK157207.1 - 7 - 8 974 91
AK177227.1 - 4 - 5 333 91
BC021352.1 - 7 - 8 846 91

View Full Product Details