The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008428.3 | NP_001008428.1 | 7 - 8 | 1050 | 80 |
GenBank mRNA | AK135785.1 | - | 7 - 8 | 1031 | 80 |
AK136200.1 | - | 7 - 8 | 1028 | 80 | |
BC085247.1 | - | 7 - 8 | 1031 | 80 |