Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cox5b
- Entrez Gene ID
-
12859
- Gene Name
- cytochrome c oxidase subunit Vb
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 36691487 - 36693388 on Build GRCm38
- Gene Symbol
- Gm11273
- Entrez Gene ID
-
100040340
- Gene Name
- cytochrome c oxidase, subunit Vb pseudogene
- Gene Aliases
-
OTTMUSG00000000426
- Chromosome Location
-
Chr.13: 21500935 - 21501452 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC023228.1 |
- |
- |
129 |
59 |
View Full Product Details