Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sema3f
- Entrez Gene ID
-
20350
- Gene Name
- sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F
- Gene Aliases
-
Sema4, Semak
- Chromosome Location
-
Chr.9: 107681499 - 107710475 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK172610.1 |
- |
|
935 |
|
Chr. - - [-] on Build |
59 |
View Full Product Details