Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d2b
Entrez Gene ID
67016
Gene Name
TBC1 domain family, member 2B
Gene Aliases
1810061M12Rik, AI480956, AV021536, AV023399, AW491493, mKIAA1055
Chromosome Location
Chr.9: 90202047 - 90270769 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194334.2 NP_919315.2 7 - 8 1645 72
GenBank mRNA AK089175.1 - 7 - 8 1611 72
AK129273.1 - 7 - 8 1631 72
AK156788.1 - 7 - 8 1632 72
BC108420.1 - 2 - 3 250 72

View Full Product Details