Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfpl1
Entrez Gene ID
81909
Gene Name
zinc finger like protein 1
Gene Aliases
1500015B20Rik
Chromosome Location
Chr.19: 6077366 - 6084939 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024231.2 NP_077193.1 4 - 5 447 135
GenBank mRNA AK005244.1 - 4 - 5 459 135
BC002119.1 - 4 - 5 447 135

View Full Product Details