The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194921.1 | NP_001181850.1 | 1 - 2 | 273 | 55 |
NM_001194923.1 | NP_001181852.1 | 1 - 2 | 273 | 55 | |
GenBank mRNA | AF349451.1 | - | 1 - 2 | 274 | 55 |
AF349453.1 | - | 1 - 2 | 274 | 55 | |
AK033657.1 | - | 1 - 2 | 274 | 55 |