Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pccb
Entrez Gene ID
66904
Gene Name
propionyl Coenzyme A carboxylase, beta polypeptide
Gene Aliases
1300012P06Rik, AI314687, R74805
Chromosome Location
Chr.9: 100982032 - 101034940 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025835.2 NP_080111.1 6 - 7 679 64
GenBank mRNA AF327060.1 - 6 - 7 667 64
AK004976.1 - 6 - 7 680 64
AK050366.1 - 6 - 7 685 64
BC026531.1 - 6 - 7 655 64
BC037082.1 - 6 - 7 681 64
BC055946.1 - 6 - 7 686 64

View Full Product Details