Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amotl2
Entrez Gene ID
56332
Gene Name
angiomotin-like 2
Gene Aliases
AW549739, C79691, Lccp, MASCOT, mKIAA0989
Chromosome Location
Chr.9: 102717294 - 102733418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019764.2 NP_062738.2 1 - 2 99 110
GenBank mRNA AK077535.1 - 1 - 2 96 110
AK141348.1 - 1 - 2 99 110
AK164279.1 - 1 - 2 168 110
AK164819.1 - 1 - 2 99 110
AK169419.1 - 1 - 2 99 110
BC035201.1 - 1 - 2 81 110

View Full Product Details