Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mllt1
Entrez Gene ID
64144
Gene Name
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 1
Gene Aliases
AA407901, BAM11, ENL, LTG19
Chromosome Location
Chr.17: 56892608 - 56935484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022328.2 NP_071723.1 3 - 4 393 82
GenBank mRNA AF298887.1 - 2 - 3 279 82
AF312858.1 - 3 - 4 290 82
AK144605.1 - 3 - 4 421 82
AK156820.1 - 3 - 4 420 82
BC053005.1 - 3 - 4 393 82

View Full Product Details