Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcta
Entrez Gene ID
102791
Gene Name
T cell leukemia translocation altered gene
Gene Aliases
9130410M22Rik, AW553637, C85065, Tctal
Chromosome Location
Chr.9: 108302958 - 108306125 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133986.1 NP_598747.1 1 - 2 227 73
GenBank mRNA AK133663.1 - - 266 73
BC019397.1 - 1 - 2 227 73

View Full Product Details