The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013598.2 | NP_038626.1 | 9 - 10 | 1062 | 98 |
GenBank mRNA | AK018777.1 | - | 9 - 10 | 1062 | 98 |
AK134301.1 | - | 9 - 10 | 1062 | 98 | |
AK165233.1 | - | 9 - 10 | 1060 | 98 | |
BC011322.1 | - | 9 - 10 | 1032 | 98 | |
M57647.1 | - | 9 - 10 | 1014 | 98 | |
S40364.1 | - | 9 - 10 | 970 | 98 | |
S40534.1 | - | 8 - 9 | 886 | 98 | |
U44725.1 | - | 9 - 10 | 1062 | 98 | |
X68989.1 | - | 8 - 9 | 865 | 98 |