Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Las1l
Entrez Gene ID
76130
Gene Name
LAS1-like (S. cerevisiae)
Gene Aliases
1810030A06Rik, 5830482G23Rik
Chromosome Location
Chr.X: 95920045 - 95956974 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152822.3 NP_690035.2 12 - 13 2276 126
GenBank mRNA AK131142.1 - 2 - 3 583 126
BC141155.1 - 12 - 13 2213 126
BM570808.1 - 1 - 2 126

View Full Product Details