The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_001463.3 | 4 - 5 | 9949 | 75 | |
NR_001570.2 | 4 - 5 | 9949 | 75 | ||
GenBank mRNA | L04961.1 | - | 4 - 5 | 9913 | 75 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | CA888102.1 | - | Chr. - - [-] on Build | 75 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | testis specific X-linked gene | Tsx | AV207184 | -44.7kb | Anti-sense | Chr. - 103409101 - 103424588 on Build GRCm38 |
Downstream | Jpx transcript, Xist activator (non-protein coding) | Jpx | 2010000I03Rik, 2510040I06Rik, Enox | 23.8kb | Anti-sense | Chr. - 103493576 - 103506425 on Build GRCm38 |