The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028576.2 | NP_082852.1 | 10 - 11 | 917 | 112 |
GenBank mRNA | AK155246.1 | - | 10 - 11 | 918 | 112 |
BC138272.1 | - | 10 - 11 | 1065 | 112 | |
BC138273.1 | - | 10 - 11 | 1065 | 112 |