The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288586.1 | NP_001275515.1 | 7 - 8 | 704 | 114 |
NM_010786.4 | NP_034916.1 | 8 - 9 | 789 | 114 | |
GenBank mRNA | AK004719.1 | - | 8 - 9 | 748 | 114 |
AK088638.1 | - | 8 - 9 | 719 | 114 | |
AK160069.1 | - | 7 - 8 | 580 | 114 | |
AK168690.1 | - | 8 - 9 | 772 | 114 | |
BC050902.1 | - | 8 - 9 | 705 | 114 | |
BC092270.1 | - | 8 - 9 | 742 | 114 | |
U47934.1 | - | 6 - 7 | 495 | 114 | |
X58876.1 | - | 8 - 9 | 696 | 114 |