Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cul2
Entrez Gene ID
71745
Gene Name
cullin 2
Gene Aliases
1300003D18Rik, 4932411N15Rik, AI327301, mKIAA4106
Chromosome Location
Chr.18: 3382975 - 3436700 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029402.3 NP_083678.2 8 - 9 906 162
GenBank mRNA AK004875.2 - 8 - 9 862 162
AK016520.1 - 8 - 9 876 162
AK028589.1 - 8 - 9 875 162
AK144693.1 - 8 - 9 879 163
AK160597.1 - 8 - 9 891 162
AK220287.1 - 8 - 9 881 162
BC026779.1 - 8 - 9 807 162
BC027428.1 - 8 - 9 869 162

View Full Product Details