Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Morn2
Entrez Gene ID
378462
Gene Name
MORN repeat containing 2
Gene Aliases
Mopt
Chromosome Location
Chr.17: 80290212 - 80305691 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC037365.1 - 1 - 2 112 68

View Full Product Details