Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppp2r5b
Entrez Gene ID
225849
Gene Name
protein phosphatase 2, regulatory subunit B', beta
Gene Aliases
B'beta, BC026670
Chromosome Location
Chr.19: 6227767 - 6235840 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198168.3 NP_937811.1 9 - 10 1547 70
GenBank mRNA AK034167.1 - 9 - 10 1566 70
AK155872.1 - 9 - 10 1540 70
AK171041.1 - 9 - 10 1518 70
AK171178.1 - 9 - 10 1548 70
AK171430.1 - 9 - 10 1555 70
BC026670.1 - 5 - 6 403 70
BC058977.1 - 9 - 10 1338 70

View Full Product Details