Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rela
Entrez Gene ID
19697
Gene Name
v-rel reticuloendotheliosis viral oncogene homolog A (avian)
Gene Aliases
p65
Chromosome Location
Chr.19: 5637440 - 5648130 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009045.4 NP_033071.1 3 - 4 531 66
GenBank mRNA AK089489.1 - 3 - 4 276 66
AK144565.1 - 3 - 4 276 66
AK154630.1 - 3 - 4 276 66
AK155321.1 - 3 - 4 542 66
AK155889.1 - 3 - 4 273 66
AK157076.1 - 3 - 4 263 66
AK158072.1 - 3 - 4 541 66
BC003818.1 - 3 - 4 290 66
BC094053.1 - 3 - 4 531 66
DQ020177.1 - 2 - 3 192 66
M61909.1 - 3 - 4 246 66

View Full Product Details