Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Batf2
- Entrez Gene ID
-
74481
- Gene Name
- basic leucine zipper transcription factor, ATF-like 2
- Gene Aliases
-
4933430F08Rik
- Chromosome Location
-
Chr.19: 6164392 - 6172476 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016990.1 |
- |
- |
108 |
62 |
View Full Product Details