Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl15
Entrez Gene ID
27395
Gene Name
mitochondrial ribosomal protein L15
Gene Aliases
HSPC145, MRP-L7, Rpml7
Chromosome Location
Chr.1: 4773198 - 4785745 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK017820.1 - 3 - 4 456 66

View Full Product Details