Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syn3
Entrez Gene ID
27204
Gene Name
synapsin III
Gene Aliases
-
Chromosome Location
Chr.10: 86048746 - 86499019 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164495.1 NP_001157967.1 2 - 3 569 65
NM_013722.3 NP_038750.2 2 - 3 569 65
GenBank mRNA AF192754.1 - 2 - 3 514 65
AF498252.1 - 1 - 2 316 65
AF498253.1 - 1 - 2 316 65
AK043854.1 - 2 - 3 570 65
AK048643.1 - 2 - 3 607 65
AK080557.1 - 2 - 3 530 65
AK082445.1 - 2 - 3 607 65
AK084032.1 - 2 - 3 555 65
AK086955.1 - - 516 65
AK087342.1 - 3 - 4 645 65
BC107397.2 - 1 - 2 435 65

View Full Product Details