Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Thap2
- Entrez Gene ID
-
66816
- Gene Name
- THAP domain containing, apoptosis associated protein 2
- Gene Aliases
-
2900040O07Rik, 9030625G08Rik, AI450385, AI649097
- Chromosome Location
-
Chr.10: 115369966 - 115384435 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK051198.1 |
- |
1 - 2 |
207 |
106 |
View Full Product Details