The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008058.2 | NP_032084.2 | 1 - 1 | 2551 | 65 |
GenBank mRNA | AK034561.1 | - | - | 369 | 65 |
BC039165.1 | - | 1 - 1 | 2237 | 65 |