Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpifc
Entrez Gene ID
270757
Gene Name
BPI fold containing family C
Gene Aliases
4732454E24, Bpil2
Chromosome Location
Chr.10: 85959340 - 86015898 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177772.4 NP_808440.2 8 - 9 819 59
GenBank mRNA AK028764.1 - 8 - 9 816 59
AK132518.1 - 8 - 9 855 59
BC112376.1 - 8 - 9 807 59

View Full Product Details