The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013523.3 | NP_038551.3 | 1 - 2 | 216 | 88 |
GenBank mRNA | AF095642.1 | - | 1 - 2 | 153 | 88 |
AK016635.1 | - | 1 - 2 | 216 | 88 | |
BC137990.1 | - | 1 - 2 | 213 | 88 | |
BC137991.1 | - | 1 - 2 | 213 | 88 |