Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thada
Entrez Gene ID
240174
Gene Name
thyroid adenoma associated
Gene Aliases
6530405K19, A930021O22, AU019899, BC052885, GITA
Chromosome Location
Chr.17: 84190055 - 84466261 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183021.3 NP_898842.2 20 - 21 3259 84
GenBank mRNA AK173258.1 - 5 - 6 769 84
EF222207.1 - 20 - 21 3167 84

View Full Product Details