The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011163.4 | NP_035293.1 | 15 - 16 | 1603 | 125 |
GenBank mRNA | AK028602.1 | - | 15 - 16 | 1603 | 125 |
BC016422.1 | - | 15 - 16 | 1566 | 125 | |
M65029.1 | - | 15 - 16 | 1555 | 125 | |
M93567.1 | - | 15 - 16 | 1545 | 125 | |
S48937.1 | - | 5 - 6 | 583 | 125 | |
X62550.1 | - | 5 - 6 | 583 | 125 |