The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030033.1 | NP_084309.1 | 6 - 7 | 630 | 84 |
GenBank mRNA | AK020340.1 | - | 7 - 8 | 691 | 84 |
AY705449.1 | - | 6 - 7 | 630 | 84 | |
DQ200908.1 | - | 8 - 9 | 807 | 84 |