Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Naa40
Entrez Gene ID
70999
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae)
Gene Aliases
4931433E08Rik, AU023197, Nat11
Chromosome Location
Chr.19: 7225668 - 7241222 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027643.1 NP_081919.1 6 - 7 770 101
GenBank mRNA AK016504.1 - 6 - 7 731 101
AK033510.1 - 6 - 7 592 101
AK033896.1 - 6 - 7 601 101
AK035418.1 - 5 - 6 1488 101
AK162625.1 - 6 - 7 604 101
AK170548.1 - 6 - 7 596 101
AK209446.1 - 1 - 2 50 101
BC020020.1 - 6 - 7 621 101
BC058212.1 - 1 - 2 429 101

View Full Product Details