The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290634.1 | NP_001277563.1 | 6 - 7 | 1103 | 81 |
NM_139297.6 | NP_647458.1 | 6 - 7 | 1293 | 81 | |
GenBank mRNA | AF424698.1 | - | 6 - 7 | 1141 | 81 |
AK033445.1 | - | 6 - 7 | 1140 | 81 | |
AK153885.1 | - | 5 - 6 | 2236 | 81 | |
BC023810.1 | - | 6 - 7 | 1144 | 81 | |
BC025585.1 | - | 6 - 7 | 1048 | 81 | |
BC026626.1 | - | 5 - 6 | 830 | 81 | |
BC061208.1 | - | 6 - 7 | 1180 | 81 |