The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010253.3 | NP_034383.1 | 5 - 6 | 454 | 115 |
GenBank mRNA | AK133522.1 | - | 5 - 6 | 470 | 115 |
BC044055.1 | - | 5 - 6 | 454 | 115 | |
Z23069.1 | - | 3 - 4 | 233 | 115 |