Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf3
Entrez Gene ID
213109
Gene Name
PHD finger protein 3
Gene Aliases
2310061N19Rik, mKIAA0244
Chromosome Location
Chr.1: 30802342 - 30873923 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081080.1 NP_001074549.1 11 - 12 3529 83
GenBank mRNA AK122230.1 - 7 - 8 980 83
BC099524.1 - 5 - 6 612 83
BC137913.1 - 11 - 12 3535 83

View Full Product Details